siRNA Details
Gene Symbol: SGSM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098509.1 | NP_001091979.1 | 4 | 551 | ||
NM_014853.2 | NP_055668.2 | 4 | 553 | |||
XM_011524101.2 | XP_011522403.1 | 5 | 629 | |||
XM_011524102.2 | XP_011522404.1 | 5 | 627 | |||
XM_011524103.2 | XP_011522405.1 | 5 | 628 | |||
XM_011524104.2 | XP_011522406.1 | 5 | 1153 | |||
XM_011524105.2 | XP_011522407.1 | 5 | 628 | |||
XM_011524106.1 | XP_011522408.1 | 4 | 451 | |||
XM_011524107.2 | XP_011522409.1 | 5 | 630 | |||
XM_011524108.1 | XP_011522410.1 | 4 | 451 | |||
XM_011524109.2 | XP_011522411.1 | 5 | 639 | |||
XM_011524110.2 | XP_011522412.1 | 5 | 639 | |||
XM_017025474.1 | XP_016880963.1 | 5 | 626 | |||
XM_017025475.1 | XP_016880964.1 | 5 | 627 | |||
GenBank mRNA | AB007857.3 | - | 4 | 551 | ||
AB275762.1 | - | 4 | 388 | |||
AB449878.1 | - | 4 | 376 | |||
AK294955.1 | - | 4 | 443 | |||
AK304263.1 | - | 4 | 349 | |||
BC039204.1 | - | 4 | 553 | 4817100 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
