siRNA Details
Gene Symbol: WDR62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083961.1 | NP_001077430.1 | 28 | 3487 | ||
NM_173636.4 | NP_775907.4 | 28 | 3472 | |||
XM_005258809.2 | XP_005258866.1 | 27 | 3376 | |||
XM_011526837.1 | XP_011525139.1 | 28 | 3472 | |||
XM_011526838.1 | XP_011525140.1 | 26 | 3238 | |||
XM_011526839.1 | XP_011525141.1 | 26 | 3136 | |||
XM_011526840.2 | XP_011525142.1 | 21 | 2482 | |||
XM_011526841.2 | XP_011525143.1 | 19 | 2103 | |||
XM_011526842.1 | XP_011525144.1 | 18 | 1935 | |||
XM_011526843.1 | XP_011525145.1 | 11 | 1262 | |||
XM_011526844.2 | XP_011525146.1 | 11 | 1259 | |||
XM_017026665.1 | XP_016882154.1 | 28 | 3487 | |||
GenBank mRNA | AL117438.1 | - | 2 | 177 | ||
AL133651.1 | - | 7 | 789 | |||
BC017261.2 | - | 10 | 1181 | |||
BC058939.1 | - | 28 | 3431 | |||
BX647726.1 | - | 26 | 4718 | |||
KU178897.1 | - | 7 | 717 | |||
KU178898.1 | - | 7 | 732 | |||
KU178899.1 | - | 5 | 483 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
