siRNA Details
Gene Symbol: WDR62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083961.1 | NP_001077430.1 | 31 | 4389 | ||
NM_173636.4 | NP_775907.4 | 31 | 4374 | |||
XM_005258809.2 | XP_005258866.1 | 30 | 4278 | |||
XM_011526837.1 | XP_011525139.1 | 31 | 4374 | |||
XM_011526838.1 | XP_011525140.1 | 29 | 4140 | |||
XM_011526839.1 | XP_011525141.1 | 29 | 4038 | |||
XM_011526840.2 | XP_011525142.1 | 24 | 3384 | |||
XM_011526841.2 | XP_011525143.1 | 22 | 3005 | |||
XM_011526842.1 | XP_011525144.1 | 21 | 2837 | |||
XM_011526843.1 | XP_011525145.1 | 14 | 2164 | |||
XM_011526844.2 | XP_011525146.1 | 14 | 2161 | |||
XM_017026665.1 | XP_016882154.1 | 31 | 4389 | |||
GenBank mRNA | AL117438.1 | - | 5 | 1079 | ||
AL133651.1 | - | 10 | 1689 | |||
BC017261.2 | - | 13 | 2083 | |||
BC058939.1 | - | 31 | 4333 | |||
BX647726.1 | - | 29 | 5620 | |||
KU178897.1 | - | 10 | 1619 | |||
KU178898.1 | - | 10 | 1634 | |||
KU178899.1 | - | 8 | 1385 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
