siRNA Details
Gene Symbol: C1orf162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300834.1 | NP_001287763.1 | 6 | 871 | ||
NM_001300835.1 | NP_001287764.1 | 5 | 851 | |||
NM_174896.3 | NP_777556.1 | 6 | 946 | |||
XM_011540712.2 | XP_011539014.1 | 7 | 6426 | |||
XM_017000323.1 | XP_016855812.1 | 7 | 2220 | |||
XM_017000324.1 | XP_016855813.1 | 6 | 6331 | |||
GenBank mRNA | AK123160.1 | - | 3 | 2225 | ||
BC017973.1 | - | 6 | 833 | 4693393 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
