siRNA Details
Gene Symbol: ILDR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199351.2 | NP_955383.1 | 2 | 247 | ||
XM_017001252.1 | XP_016856741.1 | 2 | 419 | |||
XM_017001253.1 | XP_016856742.1 | 2 | 554 | |||
XM_017001254.1 | XP_016856743.1 | 2 | 419 | |||
XM_017001255.1 | XP_016856744.1 | 2 | 419 | |||
XM_017001256.1 | XP_016856745.1 | 2 | 419 | |||
XM_017001257.1 | XP_016856746.1 | 2 | 419 | |||
XM_017001258.1 | XP_016856747.1 | 2 | 419 | |||
GenBank mRNA | AF503509.1 | - | 2 | 247 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
