siRNA Details
Gene Symbol: DQX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133637.2 | NP_598376.2 | 3 | 548 | ||
XM_011532644.1 | XP_011530946.1 | 2 | 227 | |||
XM_011532646.2 | XP_011530948.1 | 3 | 548 | |||
GenBank mRNA | BC075018.2 | - | 1 | 75 | 30915408 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
