siRNA Details
Gene Symbol: DQX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133637.2 | NP_598376.2 | 4 | 788 | ||
XM_011532644.1 | XP_011530946.1 | 3 | 467 | |||
XM_011532646.2 | XP_011530948.1 | 4 | 788 | |||
XM_017003520.1 | XP_016859009.1 | 2 | 241 | |||
GenBank mRNA | AK074337.1 | - | 4 | 695 | ||
AK092420.1 | - | 4 | 789 | |||
BC075018.2 | - | 2 | 315 | 30915408 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
