siRNA Details
Gene Symbol: NBEAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114132.1 | NP_001107604.1 | 51 | 7782 | ||
XM_005246787.3 | XP_005246844.1 | 51 | 7808 | |||
XM_005246788.2 | XP_005246845.1 | 30 | 4284 | |||
XM_006712698.3 | XP_006712761.1 | 52 | 7895 | |||
XM_006712699.3 | XP_006712762.1 | 52 | 7895 | |||
XM_011511658.2 | XP_011509960.1 | 52 | 7850 | |||
XM_011511659.2 | XP_011509961.1 | 52 | 7907 | |||
XM_011511660.2 | XP_011509962.1 | 48 | 7229 | |||
XM_011511662.2 | XP_011509964.1 | 44 | 6564 | |||
XM_017004728.1 | XP_016860217.1 | 39 | 5962 | |||
XM_017004729.1 | XP_016860218.1 | 27 | 3976 | |||
GenBank mRNA | AB053319.1 | - | 1 | 33 | ||
AK127772.1 | - | 23 | 3443 | |||
AK128636.1 | - | 21 | 2509 | |||
AY172970.1 | - | 22 | 2688 | |||
BX647133.1 | - | 15 | 1609 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
