siRNA Details
Gene Symbol: NBEAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114132.1 | NP_001107604.1 | 52 | 8014 | ||
XM_005246787.3 | XP_005246844.1 | 52 | 8040 | |||
XM_005246788.2 | XP_005246845.1 | 31 | 4516 | |||
XM_006712698.3 | XP_006712761.1 | 53 | 8127 | |||
XM_006712699.3 | XP_006712762.1 | 53 | 8127 | |||
XM_011511658.2 | XP_011509960.1 | 53 | 8082 | |||
XM_011511659.2 | XP_011509961.1 | 53 | 8139 | |||
XM_011511660.2 | XP_011509962.1 | 49 | 7461 | |||
XM_011511662.2 | XP_011509964.1 | 45 | 6796 | |||
XM_017004728.1 | XP_016860217.1 | 40 | 6194 | |||
XM_017004729.1 | XP_016860218.1 | 28 | 4208 | |||
GenBank mRNA | AB053319.1 | - | 2 | 265 | ||
AK127772.1 | - | 25 | 3612 | |||
AK128636.1 | - | 22 | 2741 | |||
AY172970.1 | - | 23 | 2920 | |||
BX647133.1 | - | 16 | 1841 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
