siRNA Details
Gene Symbol: NBEAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114132.1 | NP_001107604.1 | 53 | 8093 | ||
XM_005246787.3 | XP_005246844.1 | 53 | 8119 | |||
XM_005246788.2 | XP_005246845.1 | 32 | 4595 | |||
XM_006712698.3 | XP_006712761.1 | 54 | 8206 | |||
XM_006712699.3 | XP_006712762.1 | 54 | 8206 | |||
XM_011511658.2 | XP_011509960.1 | 54 | 8161 | |||
XM_011511659.2 | XP_011509961.1 | 54 | 8218 | |||
XM_011511660.2 | XP_011509962.1 | 50 | 7540 | |||
XM_011511662.2 | XP_011509964.1 | 46 | 6875 | |||
XM_017004728.1 | XP_016860217.1 | 41 | 6273 | |||
XM_017004729.1 | XP_016860218.1 | 29 | 4287 | |||
GenBank mRNA | AB053319.1 | - | 3 | 344 | ||
AK127772.1 | - | 26 | 3691 | |||
AK128636.1 | - | 23 | 2820 | |||
AY172970.1 | - | 24 | 2999 | |||
BX647133.1 | - | 17 | 1920 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
