siRNA Details
Gene Symbol: HERC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165136.1 | NP_001158608.1 | 3 | 568 | ||
NM_017912.3 | NP_060382.3 | 3 | 568 | |||
XM_005263083.3 | XP_005263140.1 | 3 | 765 | |||
XM_011532053.2 | XP_011530355.1 | 3 | 767 | |||
XM_017008333.1 | XP_016863822.1 | 3 | 248 | |||
GenBank mRNA | AF336798.1 | - | 3 | 568 | ||
AK092403.1 | - | 3 | 316 | |||
AK295832.1 | - | 3 | 478 | |||
AY653201.1 | - | 3 | 568 | |||
AY653202.1 | - | 3 | 568 | |||
AY653203.1 | - | 3 | 568 | |||
BC042047.1 | - | 3 | 444 | 5531519 | ||
BX647121.1 | - | 3 | 244 | |||
DA221524.1 | - | 4 | 359 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
