siRNA Details
Gene Symbol: PHIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017934.5 | NP_060404.3 | 36 | 4386 | ||
XM_005248729.4 | XP_005248786.1 | 36 | 4383 | |||
XM_011535918.2 | XP_011534220.1 | 33 | 4042 | |||
XM_017010989.1 | XP_016866478.1 | 38 | 4528 | |||
XM_017010990.1 | XP_016866479.1 | 35 | 4126 | |||
GenBank mRNA | AF310250.1 | - | 13 | 1393 | ||
AJ303102.1 | - | 36 | 4173 | |||
AK075124.1 | - | 5 | 473 | |||
AL161957.1 | - | 13 | 4240 | |||
BC008909.2 | - | 11 | 1114 | |||
BC064611.1 | - | 8 | 855 | |||
BC081569.1 | - | 14 | 1429 | |||
BC137488.1 | - | 36 | 4266 | 9021488 | ||
DQ924532.1 | - | 36 | 4160 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
