siRNA Details
Gene Symbol: PHIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017934.5 | NP_060404.3 | 37 | 4530 | ||
XM_005248729.4 | XP_005248786.1 | 37 | 4527 | |||
XM_011535918.2 | XP_011534220.1 | 34 | 4186 | |||
XM_017010989.1 | XP_016866478.1 | 39 | 4672 | |||
XM_017010990.1 | XP_016866479.1 | 36 | 4270 | |||
GenBank mRNA | AF310250.1 | - | 14 | 1537 | ||
AJ303102.1 | - | 37 | 4317 | |||
AK000712.1 | - | 10 | 1160 | |||
AK075124.1 | - | 6 | 617 | |||
AL161957.1 | - | 14 | 4384 | |||
BC008909.2 | - | 12 | 1258 | |||
BC064611.1 | - | 9 | 999 | |||
BC081569.1 | - | 15 | 1573 | |||
BC137488.1 | - | 37 | 4410 | 9021488 | ||
DQ924532.1 | - | 37 | 4304 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
