siRNA Details
Gene Symbol: TBC1D32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152730.5 | NP_689943.4 | 24 | 2821 | ||
NR_104452.1 | 27 | 3219 | ||||
XM_005266861.2 | XP_005266918.1 | 25 | 2944 | |||
XM_011535569.1 | XP_011533871.1 | 25 | 2944 | |||
XM_011535570.1 | XP_011533872.1 | 26 | 2992 | |||
XM_011535571.2 | XP_011533873.1 | 26 | 3645 | |||
XM_011535574.1 | XP_011533876.1 | 25 | 2940 | |||
XM_011535575.2 | XP_011533877.1 | 26 | 2992 | |||
XM_011535576.2 | XP_011533878.1 | 26 | 2992 | |||
XM_011535580.2 | XP_011533882.1 | 16 | 1795 | |||
XM_017010397.1 | XP_016865886.1 | 26 | 2992 | |||
XM_017010398.1 | XP_016865887.1 | 26 | 3640 | |||
XM_017010399.1 | XP_016865888.1 | 25 | 2869 | |||
XM_017010400.1 | XP_016865889.1 | 25 | 2866 | |||
XM_017010401.1 | XP_016865890.1 | 25 | 2869 | |||
XM_017010402.1 | XP_016865891.1 | 25 | 2944 | |||
XM_017010403.1 | XP_016865892.1 | 26 | 2992 | |||
XM_017010404.1 | XP_016865893.1 | 11 | 1215 | |||
GenBank mRNA | AK055461.1 | - | 8 | 766 | ||
AK091554.1 | - | 24 | 2802 | |||
AK125385.1 | - | 14 | 1654 | |||
AK126259.1 | - | 4 | 390 | |||
AK131446.1 | - | 27 | 3199 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
