siRNA Details
Gene Symbol: TBC1D32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152730.5 | NP_689943.4 | 24, 25 | 2845 | ||
XM_005266861.2 | XP_005266918.1 | 25, 26 | 2968 | |||
XM_017010397.1 | XP_016865886.1 | 26, 27 | 3016 | |||
XM_017010398.1 | XP_016865887.1 | 26, 27 | 3664 | |||
XM_017010401.1 | XP_016865890.1 | 25, 26 | 2893 | |||
XM_017010402.1 | XP_016865891.1 | 25, 26 | 2968 | |||
GenBank mRNA | AK055461.1 | - | 8, 9 | 790 | ||
AK091554.1 | - | 24, 25 | 2826 | |||
AK125385.1 | - | 14, 15 | 1678 | |||
AK126259.1 | - | 4, 5 | 414 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
