siRNA Details
Gene Symbol: TBC1D32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152730.5 | NP_689943.4 | 25 | 2895 | ||
NR_104452.1 | 29 | 3356 | ||||
XM_005266861.2 | XP_005266918.1 | 26 | 3018 | |||
XM_011535569.1 | XP_011533871.1 | 27 | 3081 | |||
XM_011535570.1 | XP_011533872.1 | 28 | 3129 | |||
XM_011535571.2 | XP_011533873.1 | 28 | 3782 | |||
XM_011535574.1 | XP_011533876.1 | 27 | 3077 | |||
XM_011535575.2 | XP_011533877.1 | 28 | 3129 | |||
XM_011535576.2 | XP_011533878.1 | 28 | 3129 | |||
XM_011535580.2 | XP_011533882.1 | 18 | 1932 | |||
XM_017010397.1 | XP_016865886.1 | 27 | 3066 | |||
XM_017010398.1 | XP_016865887.1 | 27 | 3714 | |||
XM_017010399.1 | XP_016865888.1 | 27 | 3006 | |||
XM_017010400.1 | XP_016865889.1 | 27 | 3003 | |||
XM_017010401.1 | XP_016865890.1 | 26 | 2943 | |||
XM_017010402.1 | XP_016865891.1 | 26 | 3018 | |||
XM_017010404.1 | XP_016865893.1 | 13 | 1352 | |||
GenBank mRNA | AK055461.1 | - | 9 | 840 | ||
AK091554.1 | - | 25 | 2876 | |||
AK125385.1 | - | 15 | 1728 | |||
AK126259.1 | - | 5 | 464 | |||
AK131446.1 | - | 29 | 3336 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
