siRNA Details
Gene Symbol: C7orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303470.1 | NP_001290399.1 | 9 | 1020 | ||
NM_018275.4 | NP_060745.3 | 9 | 1520 | |||
GenBank mRNA | AK001787.1 | - | 5 | 594 | ||
AK054872.1 | - | 9 | 1016 | |||
AK302342.1 | - | 7 | 868 | |||
AL136555.1 | - | 2 | 219 | |||
BC002749.2 | - | 9 | 920 | |||
BC015722.2 | - | 9 | 1478 | 3920412 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
