siRNA Details
Gene Symbol: TMEM26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178505.7 | NP_848600.2 | 4 | 850 | ||
NR_134507.1 | 4 | 850 | ||||
NR_134508.1 | 4 | 850 | ||||
XM_011539450.2 | XP_011537752.1 | 4 | 638 | |||
XM_011539451.1 | XP_011537753.1 | 4 | 638 | |||
XM_017015829.1 | XP_016871318.1 | 4 | 638 | |||
GenBank mRNA | AK125611.1 | - | 4 | 638 | ||
BC042872.1 | - | 4 | 567 | 5164050 | ||
CR749606.1 | - | 4 | 850 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
