siRNA Details
Gene Symbol: SYT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290332.1 | NP_001277261.1 | 7, 8 | 1120 | ||
NM_001290333.1 | NP_001277262.1 | 7, 8 | 1117 | |||
NM_001290334.1 | NP_001277263.1 | 7, 8 | 1101 | |||
NM_138567.4 | NP_612634.3 | 7, 8 | 1104 | |||
XM_011520455.1 | XP_011518757.1 | 7, 8 | 1622 | |||
XM_011520457.2 | XP_011518759.1 | 8, 9 | 1129 | |||
XM_017018528.1 | XP_016874017.1 | 7, 8 | 1619 | |||
XM_017018529.1 | XP_016874018.1 | 7, 8 | 981 | |||
GenBank mRNA | AK075202.1 | - | 5, 6 | 1219 | ||
AK131292.1 | - | 4, 5 | 2066 | |||
AL137708.1 | - | 1, 2 | 3082 | |||
BC144657.1 | - | 6, 7 | 1039 | |||
BC144659.1 | - | 7, 8 | 895 | |||
BC144660.1 | - | 6, 7 | 1037 | |||
BC144661.1 | - | 5, 6 | 868 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
