siRNA Details
Gene Symbol: SYT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290332.1 | NP_001277261.1 | 2 | 396 | ||
NM_001290333.1 | NP_001277262.1 | 2 | 393 | |||
NM_001290334.1 | NP_001277263.1 | 2 | 377 | |||
NM_138567.4 | NP_612634.3 | 2 | 380 | |||
XM_011520455.1 | XP_011518757.1 | 2 | 898 | |||
XM_011520456.2 | XP_011518758.1 | 2 | 898 | |||
XM_011520457.2 | XP_011518759.1 | 3 | 405 | |||
XM_017018528.1 | XP_016874017.1 | 2 | 895 | |||
XM_017018529.1 | XP_016874018.1 | 2 | 257 | |||
GenBank mRNA | AK075202.1 | - | 2 | 214 | ||
AK131292.1 | - | 2 | 898 | |||
AK293395.1 | - | 3 | 410 | |||
AL137708.1 | - | 1 | 1392 | |||
AY353087.1 | - | 1 | 76 | |||
BC144657.1 | - | 2 | 174 | |||
BC144659.1 | - | 2 | 171 | |||
BC144660.1 | - | 2 | 171 | |||
BC144661.1 | - | 2 | 171 | |||
BP225728.1 | - | 2 | 138 | |||
BX396540.2 | - | 2 | 246 | |||
BX399734.2 | - | 2 | 396 | |||
CV572958.1 | - | 2 | 166 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
