siRNA Details
Gene Symbol: SYT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290332.1 | NP_001277261.1 | 5 | 789 | ||
NM_001290333.1 | NP_001277262.1 | 5 | 786 | |||
NM_001290334.1 | NP_001277263.1 | 5 | 770 | |||
NM_138567.4 | NP_612634.3 | 5 | 773 | |||
XM_011520455.1 | XP_011518757.1 | 5 | 1291 | |||
XM_011520456.2 | XP_011518758.1 | 5 | 1291 | |||
XM_011520457.2 | XP_011518759.1 | 6 | 798 | |||
XM_017018528.1 | XP_016874017.1 | 5 | 1288 | |||
XM_017018529.1 | XP_016874018.1 | 5 | 650 | |||
GenBank mRNA | AK075202.1 | - | 4 | 748 | ||
AK131292.1 | - | 4 | 1432 | |||
AK293395.1 | - | 5 | 944 | |||
AL137708.1 | - | 1 | 2448 | |||
BC144657.1 | - | 4 | 708 | |||
BC144659.1 | - | 5 | 564 | |||
BC144661.1 | - | 4 | 705 | |||
BX396540.2 | - | 5 | 639 | |||
CV572958.1 | - | 5 | 559 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
