siRNA Details
Gene Symbol: ROBO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022370.3 | NP_071765.2 | 11 | 1907 | ||
XM_017018122.1 | XP_016873611.1 | 10 | 1672 | |||
GenBank mRNA | AK291045.1 | - | 11 | 1841 | ||
AY509035.1 | - | 11 | 1930 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
