siRNA Details
Gene Symbol: SPATA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144644.3 | NP_653245.2 | 4, 5 | 800 | ||
GenBank mRNA | AY040204.3 | - | 4, 5 | 800 | ||
BC021731.2 | - | 4, 5 | 701 | 5269042 | ||
BC039342.1 | - | 3, 4 | 1768 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
