siRNA Details
Gene Symbol: ZSCAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182572.3 | NP_872378.3 | 5 | 780 | ||
XM_006723149.2 | XP_006723212.1 | 4 | 2393 | |||
GenBank mRNA | AK091098.1 | - | 5 | 780 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
