siRNA Details
Gene Symbol: ZSCAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182572.3 | NP_872378.3 | 3 | 384 | ||
XM_006723149.2 | XP_006723212.1 | 2 | 1997 | |||
XM_017026609.1 | XP_016882098.1 | 3 | 943 | |||
XM_017026610.1 | XP_016882099.1 | 3 | 384 | |||
GenBank mRNA | AK091098.1 | - | 3 | 384 | ||
AY280370.1 | - | 3 | 344 | |||
BC048099.1 | - | 3 | 344 | |||
BC107729.1 | - | 4 | 485 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
