siRNA Details
Gene Symbol: SREK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077199.2 | NP_001070667.1 | 9 | 1576 | ||
NM_001270492.1 | NP_001257421.1 | 10 | 1700 | |||
NM_001323527.1 | NP_001310456.1 | 10 | 1700 | |||
NM_001323529.1 | NP_001310458.1 | 9 | 1576 | |||
NM_001323533.1 | NP_001310462.1 | 9 | 1576 | |||
NM_001323534.1 | NP_001310463.1 | 10 | 1591 | |||
NM_001323535.1 | NP_001310464.1 | 9 | 1467 | |||
NM_139168.3 | NP_631907.1 | 10 | 1729 | |||
XM_011543171.2 | XP_011541473.1 | 9 | 1801 | |||
XM_017009056.1 | XP_016864545.1 | 9 | 1361 | |||
GenBank mRNA | AB209694.1 | - | 2 | 6870 | ||
AF459094.1 | - | 9 | 1410 | |||
AK091758.1 | - | 9 | 1459 | |||
AK125893.1 | - | 2 | 1261 | |||
AK308287.1 | - | 10 | 1603 | |||
BC067770.1 | - | 10 | 1730 | 4827589 | ||
BC112343.1 | - | 7 | 1098 | 40008558 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
