siRNA Details
Gene Symbol: SREK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077199.2 | NP_001070667.1 | 11, 12 | 1887 | ||
NM_001270492.1 | NP_001257421.1 | 12, 13 | 2008 | |||
NM_001323527.1 | NP_001310456.1 | 12, 13 | 2011 | |||
NM_001323529.1 | NP_001310458.1 | 11, 12 | 1884 | |||
NM_001323534.1 | NP_001310463.1 | 12, 13 | 1902 | |||
NM_001323535.1 | NP_001310464.1 | 11, 12 | 1775 | |||
NM_139168.3 | NP_631907.1 | 12, 13 | 2040 | |||
XM_011543171.2 | XP_011541473.1 | 11, 12 | 2112 | |||
XM_017009055.1 | XP_016864544.1 | 10, 11 | 1421 | |||
XM_017009056.1 | XP_016864545.1 | 11, 12 | 1672 | |||
GenBank mRNA | AF459094.1 | - | 11, 12 | 1721 | ||
AK091758.1 | - | 11, 12 | 1769 | |||
AK125893.1 | - | 4, 5 | 1572 | |||
AK308287.1 | - | 12, 13 | 1910 | |||
BC067770.1 | - | 12, 13 | 2041 | 4827589 | ||
BC112343.1 | - | 9, 10 | 1409 | 40008558 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
