siRNA Details
Gene Symbol: CCDC174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016474.4 | NP_057558.3 | 7 | 725 | ||
NR_135523.1 | 7 | 725 | ||||
XM_017006554.1 | XP_016862043.1 | 7 | 788 | |||
XM_017006555.1 | XP_016862044.1 | 7 | 786 | |||
GenBank mRNA | AF151046.1 | - | 7 | 691 | ||
AK309525.1 | - | 7 | 698 | |||
BC013999.2 | - | 7 | 701 | 3936914 | ||
BC033897.1 | - | 7 | 712 | 4812977 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
