siRNA Details
Gene Symbol: VAMP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145149.2 | NP_001138621.1 | 3 | 385 | ||
NM_001185183.1 | NP_001172112.1 | 4 | 508 | |||
NM_005638.5 | NP_005629.1 | 4 | 508 | |||
NR_033714.1 | 3 | 450 | ||||
NR_033715.1 | 4 | 443 | ||||
XM_011531188.1 | XP_011529490.1 | 4 | 487 | |||
XM_017029760.1 | XP_016885249.1 | 4 | 491 | |||
GenBank mRNA | AJ295938.1 | - | 4 | 420 | ||
AJ549301.2 | - | 3 | 297 | |||
AK222794.1 | - | 4 | 409 | |||
AK293525.1 | - | 3 | 368 | |||
AK295605.1 | - | 4 | 368 | |||
AK297385.1 | - | 3 | 372 | |||
AK312658.1 | - | 4 | 473 | |||
BC056141.1 | - | 4 | 508 | 6503665 | ||
DC341378.1 | - | 4 | 439 | |||
X92396.1 | - | 4 | 444 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
