siRNA Details
Gene Symbol: CLDN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199328.2 | NP_955360.1 | 1 | 1194 | ||
GenBank mRNA | AK022269.1 | - | 1 | 1112 | ||
AL049977.1 | - | 1 | 128 | |||
AY358707.1 | - | 1 | 1064 | |||
BC020866.1 | - | 2 | 853 | 4594155 | ||
BC058004.1 | - | 1 | 1043 | 4606880 | ||
GQ472207.1 | - | 2 | 853 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
