siRNA Details
Gene Symbol: SSBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009955.3 | NP_001009955.1 | 17 | 1634 | ||
NM_018070.4 | NP_060540.2 | 17 | 1655 | |||
NM_145716.3 | NP_663768.1 | 18 | 1715 | |||
XM_006710545.3 | XP_006710608.1 | 16 | 1214 | |||
XM_017000897.1 | XP_016856386.1 | 18 | 1384 | |||
XM_017000898.1 | XP_016856387.1 | 15 | 1611 | |||
XM_017000899.1 | XP_016856388.1 | 17 | 1315 | |||
GenBank mRNA | AF086085.1 | - | 4 | 353 | ||
AF500116.1 | - | 17 | 1483 | |||
AK001217.1 | - | 14 | 1063 | |||
AK093975.1 | - | 14 | 1375 | |||
AK289474.1 | - | 17 | 1256 | |||
BC003605.1 | - | 17 | 1294 | 3546126 | ||
BC004335.1 | - | 11 | 812 | |||
BC066365.1 | - | 18 | 1459 | 6169677 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
