siRNA Details
Gene Symbol: LDLRAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015627.2 | NP_056442.2 | 5 | 585 | ||
XM_006710559.3 | XP_006710622.1 | 5 | 582 | |||
XM_006710560.3 | XP_006710623.1 | 5 | 582 | |||
XM_006710561.3 | XP_006710624.1 | 5 | 582 | |||
XM_011541209.2 | XP_011539511.1 | 5 | 582 | |||
XM_011541210.2 | XP_011539512.1 | 5 | 582 | |||
XM_011541211.2 | XP_011539513.1 | 5 | 582 | |||
XM_011541212.2 | XP_011539514.1 | 5 | 582 | |||
XM_017000994.1 | XP_016856483.1 | 5 | 1527 | |||
GenBank mRNA | AK024397.1 | - | 4 | 943 | ||
AK091219.1 | - | 5 | 590 | |||
AL117654.1 | - | 5 | 492 | |||
BC029770.1 | - | 5 | 495 | 5197824 | ||
DA930479.1 | - | 2 | 57 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
