siRNA Details
Gene Symbol: LDLRAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015627.2 | NP_056442.2 | 8, 9 | 904 | ||
XM_011541209.2 | XP_011539511.1 | 8, 9 | 898 | |||
XM_017000994.1 | XP_016856483.1 | 8, 9 | 1846 | |||
GenBank mRNA | AK024397.1 | - | 6, 7 | 2643 | ||
AK091219.1 | - | 8, 9 | 909 | |||
AK309850.1 | - | 4, 5 | 443 | |||
AK309853.1 | - | 3, 4 | 521 | |||
AL117654.1 | - | 8, 9 | 811 | |||
BC029770.1 | - | 8, 9 | 814 | 5197824 | ||
DA930479.1 | - | 5, 6 | 376 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
