siRNA Details
Gene Symbol: ZNF782 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001662.1 | NP_001001662.1 | 6 | 2176 | ||
XM_005251742.4 | XP_005251799.1 | 5 | 2195 | |||
XM_005251744.3 | XP_005251801.1 | 4 | 2136 | |||
XM_011518315.2 | XP_011516617.1 | 6 | 3387 | |||
XM_011518318.2 | XP_011516620.1 | 6 | 2374 | |||
XM_011518320.2 | XP_011516622.1 | 5 | 2011 | |||
XM_017014343.1 | XP_016869832.1 | 5 | 3563 | |||
XM_017014344.1 | XP_016869833.1 | 4 | 1908 | |||
GenBank mRNA | AK131468.1 | - | 6 | 2176 | ||
AK302460.1 | - | 3 | 1860 | |||
BC137073.1 | - | 6 | 2173 | 9021070 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
