siRNA Details
Gene Symbol: NUCKS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022731.4 | NP_073568.2 | 7 | 972 | ||
XM_005245453.1 | XP_005245510.1 | 7 | 945 | |||
GenBank mRNA | AB049824.1 | - | 7 | 903 | ||
AJ012584.1 | - | 7 | 799 | |||
AK025133.1 | - | 7 | 767 | |||
AK225228.1 | - | 7 | 767 | |||
AY823399.1 | - | 6 | 622 | |||
BC000805.1 | - | 7 | 893 | 3452665 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
