siRNA Details
Gene Symbol: VPS53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128159.2 | NP_001121631.1 | 18 | 2131 | ||
NM_018289.3 | NP_060759.2 | 17 | 2044 | |||
XM_011523953.2 | XP_011522255.1 | 19 | 2219 | |||
XM_017024817.1 | XP_016880306.1 | 15 | 2177 | |||
XM_017024818.1 | XP_016880307.1 | 14 | 2372 | |||
GenBank mRNA | AF246287.1 | - | 18 | 2042 | ||
AK001841.1 | - | 8 | 1025 | |||
AK091388.1 | - | 13 | 1754 | |||
AK092532.1 | - | 19 | 2229 | |||
AK290343.1 | - | 17 | 2024 | |||
AY444797.1 | - | 18 | 2039 | |||
BC006116.1 | - | 17 | 1956 | 3504453 | ||
EU021218.1 | - | 18 | 1984 | |||
EU434648.1 | - | 17 | 1787 | |||
EU434649.1 | - | 11 | 1153 | |||
EU434650.1 | - | 17 | 1897 |
Gene Symbol: LOC100129974 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128440.1 | - | 1 | 2146 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
