siRNA Details
Gene Symbol: VPS53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128159.2 | NP_001121631.1 | 16 | 1904 | ||
NM_018289.3 | NP_060759.2 | 15 | 1817 | |||
XM_011523953.2 | XP_011522255.1 | 17 | 1992 | |||
XM_017024817.1 | XP_016880306.1 | 13 | 1950 | |||
XM_017024818.1 | XP_016880307.1 | 12 | 2145 | |||
GenBank mRNA | AF246287.1 | - | 16 | 1812 | ||
AF318335.1 | - | 16 | 1875 | |||
AK001841.1 | - | 6 | 798 | |||
AK091388.1 | - | 11 | 1527 | |||
AK092532.1 | - | 17 | 2002 | |||
AK290343.1 | - | 15 | 1797 | |||
AY444797.1 | - | 16 | 1812 | |||
BC006116.1 | - | 15 | 1729 | 3504453 | ||
EU021218.1 | - | 16 | 1757 | |||
EU434649.1 | - | 9 | 926 | |||
EU434650.1 | - | 15 | 1670 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
