siRNA Details
Gene Symbol: VPS53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128159.2 | NP_001121631.1 | 14 | 1639 | ||
NM_018289.3 | NP_060759.2 | 13 | 1552 | |||
XM_011523953.2 | XP_011522255.1 | 15 | 1727 | |||
XM_017024817.1 | XP_016880306.1 | 11 | 1685 | |||
XM_017024818.1 | XP_016880307.1 | 10 | 1880 | |||
GenBank mRNA | AF246287.1 | - | 14 | 1547 | ||
AF318335.1 | - | 14 | 1609 | |||
AK001841.1 | - | 4 | 533 | |||
AK091388.1 | - | 9 | 1262 | |||
AK092532.1 | - | 15 | 1737 | |||
AK290343.1 | - | 13 | 1532 | |||
AY444797.1 | - | 14 | 1547 | |||
BC006116.1 | - | 13 | 1464 | 3504453 | ||
EU021218.1 | - | 14 | 1492 | |||
EU434648.1 | - | 13 | 1348 | |||
EU434649.1 | - | 7 | 661 | |||
EU434650.1 | - | 13 | 1405 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
