siRNA Details
Gene Symbol: CCDC155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144688.4 | NP_653289.3 | 14, 15 | 1347 | ||
XM_005258549.3 | XP_005258606.1 | 14, 15 | 1347 | |||
XM_011526488.2 | XP_011524790.1 | 14, 15 | 1347 | |||
XM_011526489.2 | XP_011524791.1 | 14, 15 | 3247 | |||
XM_011526494.2 | XP_011524796.1 | 15, 16 | 1320 | |||
XM_011526495.2 | XP_011524797.1 | 15, 16 | 1356 | |||
XM_011526496.2 | XP_011524798.1 | 13, 14 | 1208 | |||
XM_011526497.2 | XP_011524799.1 | 14, 15 | 1306 | |||
XM_017026333.1 | XP_016881822.1 | 13, 14 | 1480 | |||
GenBank mRNA | AK057220.1 | - | 14, 15 | 1316 | ||
BC029811.1 | - | 14, 15 | 1266 | 5171712 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
