siRNA Details
Gene Symbol: CCDC155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144688.4 | NP_653289.3 | 4 | 515 | ||
XM_005258549.3 | XP_005258606.1 | 4 | 515 | |||
XM_011526488.2 | XP_011524790.1 | 4 | 515 | |||
XM_011526489.2 | XP_011524791.1 | 4 | 2415 | |||
XM_011526490.2 | XP_011524792.1 | 4 | 515 | |||
XM_011526494.2 | XP_011524796.1 | 5 | 488 | |||
XM_011526495.2 | XP_011524797.1 | 5 | 524 | |||
XM_011526496.2 | XP_011524798.1 | 3 | 376 | |||
XM_011526497.2 | XP_011524799.1 | 4 | 474 | |||
XM_017026333.1 | XP_016881822.1 | 3 | 648 | |||
GenBank mRNA | AK057220.1 | - | 4 | 484 | ||
AK097696.1 | - | 5 | 1228 | |||
BC029811.1 | - | 4 | 434 | 5171712 | ||
DB024870.1 | - | 4 | 515 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
