siRNA Details
Gene Symbol: BCL6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181844.3 | NP_862827.1 | 9 | 1832 | ||
GenBank mRNA | AB076580.1 | - | 9 | 1812 | ||
AK122893.1 | - | 9 | 1826 | |||
AK292878.1 | - | 9 | 1815 | |||
AK293102.1 | - | 9 | 1829 | |||
BC059404.1 | - | 9 | 1831 | 30342006 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
