siRNA Details
Gene Symbol: BCL6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181844.3 | NP_862827.1 | 2 | 118 | ||
GenBank mRNA | AB076580.1 | - | 2 | 98 | ||
AK122893.1 | - | 2 | 114 | |||
AK292878.1 | - | 2 | 101 | |||
AK293102.1 | - | 2 | 117 | |||
BC059404.1 | - | 2 | 119 | 30342006 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
