siRNA Details
Gene Symbol: LRRTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178839.4 | NP_849161.2 | 2 | 2017 | ||
XM_017003986.1 | XP_016859475.1 | 2 | 2549 | |||
XM_017003987.1 | XP_016859476.1 | 2 | 2397 | |||
GenBank mRNA | AK056644.1 | - | 2 | 2048 | ||
AK225779.1 | - | 2 | 2404 | |||
AK290512.1 | - | 2 | 2267 | |||
AY358310.1 | - | 2 | 2466 | |||
BC045113.1 | - | 2 | 2017 | 5200499 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
