siRNA Details
Gene Symbol: NBEAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114132.1 | NP_001107604.1 | 5, 6 | 725 | ||
XM_005246787.3 | XP_005246844.1 | 5, 6 | 751 | |||
XM_006712698.3 | XP_006712761.1 | 5, 6 | 751 | |||
XM_006712699.3 | XP_006712762.1 | 5, 6 | 751 | |||
XM_011511658.2 | XP_011509960.1 | 5, 6 | 706 | |||
XM_011511659.2 | XP_011509961.1 | 5, 6 | 763 | |||
XM_011511663.2 | XP_011509965.1 | 5, 6 | 741 | |||
GenBank mRNA | AK130065.1 | - | 2, 3 | 128 | ||
AW809287.1 | - | 17 | ||||
BC132950.1 | - | 5, 6 | 679 | |||
BC137214.1 | - | 5, 6 | 679 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
