siRNA Details
Gene Symbol: NBEAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114132.1 | NP_001107604.1 | 6 | 798 | ||
XM_005246787.3 | XP_005246844.1 | 6 | 824 | |||
XM_006712698.3 | XP_006712761.1 | 6 | 824 | |||
XM_006712699.3 | XP_006712762.1 | 6 | 824 | |||
XM_011511658.2 | XP_011509960.1 | 6 | 779 | |||
XM_011511659.2 | XP_011509961.1 | 6 | 836 | |||
XM_011511660.2 | XP_011509962.1 | 2 | 158 | |||
XM_011511663.2 | XP_011509965.1 | 6 | 814 | |||
GenBank mRNA | AK130065.1 | - | 3 | 201 | ||
AW809287.1 | - | 1 | 90 | |||
BC132950.1 | - | 6 | 752 | |||
BC137214.1 | - | 6 | 752 | |||
BC144532.1 | - | 5 | 670 | |||
BC144533.1 | - | 6 | 748 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
