siRNA Details
Gene Symbol: C4orf36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144645.3 | NP_653246.1 | 4, 5 | 678 | ||
GenBank mRNA | BC016746.1 | - | 4, 5 | 654 | 4827131 | |
BX090513.1 | - | 3, 4 | 419 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
