siRNA Details
Gene Symbol: RBM47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098634.1 | NP_001092104.1 | 7 | 2360 | ||
NM_019027.3 | NP_061900.2 | 5 | 1704 | |||
XM_005248103.3 | XP_005248160.1 | 7 | 2048 | |||
XM_005248107.3 | XP_005248164.1 | 6 | 2047 | |||
XM_005248108.3 | XP_005248165.1 | 6 | 2197 | |||
XM_005248109.4 | XP_005248166.1 | 6 | 3053 | |||
XM_011513704.2 | XP_011512006.1 | 8 | 2456 | |||
XM_011513707.2 | XP_011512009.1 | 7 | 2201 | |||
XM_011513708.2 | XP_011512010.1 | 7 | 2030 | |||
XM_017008304.1 | XP_016863793.1 | 7 | 4352 | |||
XM_017008305.1 | XP_016863794.1 | 9 | 2584 | |||
XM_017008306.1 | XP_016863795.1 | 8 | 2481 | |||
XM_017008307.1 | XP_016863796.1 | 7 | 2096 | |||
XM_017008308.1 | XP_016863797.1 | 6 | 2273 | |||
XM_017008309.1 | XP_016863798.1 | 7 | 2045 | |||
XM_017008310.1 | XP_016863799.1 | 6 | 1841 | |||
GenBank mRNA | AF262323.1 | - | 6 | 1872 | ||
AK000280.1 | - | 5 | 1682 | |||
AK304206.1 | - | 6 | 1795 | |||
BC071585.1 | - | 7 | 2596 | |||
BC126261.1 | - | 5 | 1805 | 8991977 | ||
BC143942.1 | - | 5 | 1805 | 9052460 | ||
BX648404.1 | - | 5 | 382 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
