siRNA Details
Gene Symbol: RBM47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098634.1 | NP_001092104.1 | 4 | 834 | ||
NM_019027.3 | NP_061900.2 | 3 | 385 | |||
XM_005248103.3 | XP_005248160.1 | 4 | 522 | |||
XM_005248107.3 | XP_005248164.1 | 3 | 521 | |||
XM_005248108.3 | XP_005248165.1 | 3 | 671 | |||
XM_005248109.4 | XP_005248166.1 | 3 | 1527 | |||
XM_011513704.2 | XP_011512006.1 | 5 | 930 | |||
XM_011513707.2 | XP_011512009.1 | 4 | 675 | |||
XM_011513708.2 | XP_011512010.1 | 4 | 504 | |||
XM_017008304.1 | XP_016863793.1 | 4 | 2826 | |||
XM_017008305.1 | XP_016863794.1 | 6 | 1058 | |||
XM_017008306.1 | XP_016863795.1 | 5 | 955 | |||
XM_017008307.1 | XP_016863796.1 | 4 | 570 | |||
XM_017008308.1 | XP_016863797.1 | 3 | 747 | |||
XM_017008309.1 | XP_016863798.1 | 4 | 522 | |||
XM_017008310.1 | XP_016863799.1 | 4 | 522 | |||
GenBank mRNA | AF262323.1 | - | 3 | 346 | ||
AK000280.1 | - | 3 | 363 | |||
AK304206.1 | - | 3 | 269 | |||
BC034402.1 | - | 4 | 812 | |||
BC071585.1 | - | 4 | 832 | |||
BC126261.1 | - | 2 | 279 | 8991977 | ||
BC143942.1 | - | 2 | 279 | 9052460 | ||
BP332501.1 | - | 3 | 385 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
