siRNA Details
Gene Symbol: PRRT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207351.4 | NP_997234.3 | 4 | 2356 | ||
NR_134900.1 | 4 | 2356 | ||||
XM_011533628.2 | XP_011531930.1 | 4 | 2434 | |||
XM_011533629.2 | XP_011531931.1 | 4 | 2414 | |||
XM_017006256.1 | XP_016861745.1 | 3 | 4221 | |||
GenBank mRNA | BC040508.1 | - | 4 | 2333 | 5300209 | |
BC089447.1 | - | 4 | 2315 | 30563721 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
