siRNA Details
Gene Symbol: SHROOM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020859.3 | NP_065910.3 | 7, 8 | 5670 | ||
XM_005263163.4 | XP_005263220.1 | 6, 7 | 10124 | |||
XM_005263164.4 | XP_005263221.1 | 6, 7 | 4682 | |||
XM_005263165.4 | XP_005263222.1 | 6, 7 | 6108 | |||
XM_006714282.3 | XP_006714345.1 | 7, 8 | 4825 | |||
XM_011532157.2 | XP_011530459.2 | 6, 7 | 5512 | |||
XM_011532158.2 | XP_011530460.1 | 8, 9 | 5471 | |||
XM_011532159.1 | XP_011530461.1 | 7, 8 | 5660 | |||
XM_011532160.1 | XP_011530462.1 | 5, 6 | 4672 | |||
XM_011532161.2 | XP_011530463.1 | 6, 7 | 4850 | |||
XM_017008488.1 | XP_016863977.1 | 7, 8 | 4828 | |||
XM_017008489.1 | XP_016863978.1 | 7, 8 | 4729 | |||
GenBank mRNA | AB040914.1 | - | 3, 4 | 2869 | ||
AB055660.1 | - | 7, 8 | 5671 | |||
AK027744.1 | - | 4, 5 | 607 | |||
AK128677.1 | - | 3, 4 | 4172 | |||
BC007291.1 | - | 3, 4 | 1031 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
